3,5-Difluoro-2-hydroxyphenylboronic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (3,5-Difluoro-2-hydroxyphenyl)boronic acid

PC412053-1G
APOSPC412053-1GEA 201 GBP
APOSPC412053-1G
3,5-Difluoro-2-hydroxyphenylboronic acid
3,5-Difluoro-4-hydroxyphenylboronic acid
Formula: C₆H₅BF₂O₃
MDL Number: MFCD03095356
CAS Number: 1150114-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR