2-Methyl-4-(2,2,2-trifluoroethoxy)aniline hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC411321-1G PC411321-5G
APOSPC411321-1GEA 29.4 GBP
APOSPC411321-1G APOSPC411321-5G
2-Methyl-4-(2,2,2-trifluoroethoxy)aniline hydrochloride
2-Methyl-4-(2,2,2-trifluoroethoxy)aniline hydrochloride
Formula: C₉H₁₀F₃NO·ClH
MW: 241.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25371225
CAS Number: 1431962-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR