trans-3-Fluorocyclobutane-1-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: trans-3-Fluorocyclobutanecarboxylic acid

PC411270-1G
APOSPC411270-1GEA 873 GBP
APOSPC411270-1G
trans-3-Fluorocyclobutane-1-carboxylic acid
trans-3-Fluorocyclobutane-1-carboxylic acid
Formula: C₅H₇FO₂
MW: 118.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16661184
CAS Number: 123812-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR