2-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC410481-1G
APOSPC410481-1GEA 468 GBP
APOSPC410481-1G
2-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formula: C₈H₈BrF₃N₂O₂
MW: 301.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04970065
CAS Number: 1006473-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR