3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC410397-1G
APOSPC410397-1GEA 373 GBP
APOSPC410397-1G
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
Formula: C₁₀H₁₆F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD06805085
CAS Number: 1006340-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR