3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC410281-1G
APOSPC410281-1GEA 468 GBP
APOSPC410281-1G
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formula: C₁₀H₁₀ClF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419752
CAS Number: 1001518-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR