1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC410264-1G
APOSPC410264-1GEA 132 GBP
APOSPC410264-1G
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Formula: C₆H₇F₃N₄O
MW: 208.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419674
CAS Number: 1001518-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR