1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC408443-250MG PC408443-1G PC408443-5G
APOSPC408443-250MGEA 158 GBP
APOSPC408443-250MG APOSPC408443-1G APOSPC408443-5G
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde 95%
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde
Formula: C₆H₅F₃N₂O
MDL Number: MFCD28403393
CAS Number: 1557546-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR