1-(4-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC408321-500MG
APOSPC408321-500MGEA 311 GBP
APOSPC408321-500MG
1-(4-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride
1-(4-Fluoro-benzyl)-piperazine-2-carboxylic acid hydrochloride
Formula: C₁₂H₁₅FN₂O₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD18837465
CAS Number: 1289384-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR