4-Fluoro-2-methoxybenzyl alcohol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (4-Fluoro-2-methoxyphenyl)methanol

PC408257-1G PC408257-5G PC408257-10G
APOSPC408257-1GEA 32.6 GBP
APOSPC408257-1G APOSPC408257-5G APOSPC408257-10G
4-Fluoro-2-methoxybenzyl alcohol 98%
4-Fluoro-2-methoxybenzyl alcohol
Formula: C₈H₉FO₂
Boiling Pt: 110…112 °C/4mmHg
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD04116338
CAS Number: 157068-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR