Dimethyl difluoromalonate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC408111-1G PC408111-25G PC408111-5G
APOSPC408111-1GEA 23.2 GBP
APOSPC408111-1G APOSPC408111-25G APOSPC408111-5G
Dimethyl difluoromalonate 98%
Dimethyl difluoromalonate
Formula: C₅H₆F₂O₄
Boiling Pt: 167…169 °C
Density: 1.3059 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00516931
CAS Number: 379-95-3
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR