α,2-Dibromo-5-fluorotoluene 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-Bromo-2-(bromomethyl)-4-fluorobenzene, 2-Bromo-5-fluorobenzyl bromide

PC4075-100G PC4075-25G PC4075-5G
APOSPC4075-100GEA 120 GBP
APOSPC4075-100G APOSPC4075-25G APOSPC4075-5G
α,2-Dibromo-5-fluorotoluene 98+%
α,2-Dibromo-5-fluorotoluene
Formula: C₇H₅Br₂F
Boiling Pt: 89…90 °C/1mmHg
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR