4-Bromo-8-fluoroquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC407011-1G PC407011-5G
APOSPC407011-1GEA 59.9 GBP
APOSPC407011-1G APOSPC407011-5G
4-Bromo-8-fluoroquinoline
4-Bromo-8-fluoroquinoline
Formula: C₉H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD07700231
CAS Number: 927800-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR