N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N,N-Bis(trifluoromethylsulphonyl)-2-pyridylamine, 2-Pyridyltriflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]pyridine, 1,1,1-Trifluoro-N-(pyridin-2-yl)-N-((trifluoromethyl)sulphonyl)methanesulphonamide

PC4064-5G PC4064-10G PC4064-1G
APOSPC4064-5GEA 24.2 GBP
APOSPC4064-5G APOSPC4064-10G APOSPC4064-1G
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
N-(2-Pyridyl)bis(trifluoromethanesulphonimide)

Triflating reagent.

Formula: C₇H₄F₆N₂O₄S₂
Boiling Pt: 80…90 °C/0.25mmHg
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191834
CAS Number: 145100-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR