4-(Pentafluorothio)benzenesulphonyl chloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC405657-1G PC405657-5G PC405657-10G
APOSPC405657-1GEA 247 GBP
APOSPC405657-1G APOSPC405657-5G APOSPC405657-10G
4-(Pentafluorothio)benzenesulphonyl chloride 97%
4-(Pentafluorothio)benzenesulphonyl chloride
Formula: C₆H₄ClF₅O₂S₂
MW: 302.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652450
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR