4-Amino-3-bromo-6-fluoroquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC404608-1G
APOSPC404608-1GEA 368 GBP
APOSPC404608-1G
4-Amino-3-bromo-6-fluoroquinoline
4-Amino-3-bromo-6-fluoroquinoline
Formula: C₉H₆BrFN₂
MW: 241.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13619792
CAS Number: 1065088-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR