2-Chloro-3-(chloromethyl)-7-fluoroquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC404555-1G
APOSPC404555-1GEA 296 GBP
APOSPC404555-1G
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
Formula: C₁₀H₆Cl₂FN
Storage Temperature: Ambient
CAS Number: 948291-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR