4-(3-Methoxy-5-trifluoromethyl-benzyl)-piperidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC404517-1G
APOSPC404517-1GEA 350 GBP
APOSPC404517-1G
4-(3-Methoxy-5-trifluoromethyl-benzyl)-piperidine
4-(3-Methoxy-5-trifluoromethyl-benzyl)-piperidine
Formula: C₁₄H₁₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD05863576
CAS Number: 794500-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR