2-Chloro-7-fluoro-3-methylquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC404501-1G
APOSPC404501-1GEA 266 GBP
APOSPC404501-1G
2-Chloro-7-fluoro-3-methylquinoline
2-Chloro-7-fluoro-3-methylquinoline
Formula: C₁₀H₇ClFN
Storage Temperature: Ambient
CAS Number: 132118-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR