2-Fluoro-4-nitroanisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC3882-100G PC3882-25G PC3882-5G
APOSPC3882-100GEA 42 GBP
APOSPC3882-100G APOSPC3882-25G APOSPC3882-5G
2-Fluoro-4-nitroanisole 98%
2-Fluoro-4-nitroanisole
Formula: C₇H₆FNO₃
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00061095
CAS Number: 455-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR