2-Fluoro-5-nitroaniline 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC3860-100G PC3860-25G PC3860-5G
APOSPC3860-100GEA 39.9 GBP
APOSPC3860-100G APOSPC3860-25G APOSPC3860-5G
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
Melting Pt: 99…101 °C
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR