Difluoroacetic anhydride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC3791-25G PC3791-5G PC3791-100G
APOSPC3791-25GEA 42 GBP
APOSPC3791-25G APOSPC3791-5G APOSPC3791-100G
Difluoroacetic anhydride 97%
Difluoroacetic anhydride
Formula: C₄H₂F₄O₃
Boiling Pt: 125…127 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093315
CAS Number: 401-67-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR