5-Fluoroindole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 5-Fluoro-1H-indole

PC3758-5G PC3758-1G
APOSPC3758-5GEA 22.1 GBP
APOSPC3758-5G APOSPC3758-1G
5-Fluoroindole 98%
5-Fluoroindole
Formula: C₈H₆FN
Melting Pt: 45…48 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR