2-Vinylhexafluoroisopropanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC3737-5G
APOSPC3737-5GEA 172 GBP
APOSPC3737-5G
2-Vinylhexafluoroisopropanol
2-Vinylhexafluoroisopropanol
Formula: C₅H₄F₆O
Boiling Pt: 73…74 °C
Storage Temperature: Ambient
MDL Number: MFCD04038371
CAS Number: 19701-19-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR