2-Chloro-5-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC3355-1G PC3355-25G PC3355-5G
APOSPC3355-1GEA 15.8 GBP
APOSPC3355-1G APOSPC3355-25G APOSPC3355-5G
2-Chloro-5-(trifluoromethyl)benzyl bromide 98%
2-Chloro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrClF₃
Boiling Pt: 80 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00236216
CAS Number: 237761-77-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR