2-(2-Fluoroethoxy)-5-methylbenzoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC32902-100MG
APOSPC32902-100MGEA 361 GBP
APOSPC32902-100MG
2-(2-Fluoroethoxy)-5-methylbenzoic acid 95%
2-(2-Fluoroethoxy)-5-methylbenzoic acid
Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20661084
CAS Number: 1269797-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR