Ethyl-3-(trifluoromethyl)crotonate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(Trifluoromethyl)crotonic acid ethyl ester

PC3288C-25G PC3288C-5G
APOSPC3288C-25GEA 289 GBP
APOSPC3288C-25G APOSPC3288C-5G
Ethyl-3-(trifluoromethyl)crotonate 97%
Ethyl-3-(trifluoromethyl)crotonate
Formula: C₇H₉F₃O₂
MW: 182.14 g/mol
Boiling Pt: 129…130 °C
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00040846
CAS Number: 24490-03-7
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR