Difluoro(6-methylpyridin-2-yl)acetic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC32862-250MG PC32862-500MG
APOSPC32862-250MGEA 237 GBP
APOSPC32862-250MG APOSPC32862-500MG
Difluoro(6-methylpyridin-2-yl)acetic acid
Difluoro(6-methylpyridin-2-yl)acetic acid
Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD12827869
CAS Number: 1215353-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR