2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: β-Bromo-2-methoxy-α,α,β,β-tetraflurophenetole, 1-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-2-methoxybenzene

PC32761-100G PC32761-25G PC32761-5G
APOSPC32761-100GEA 537 GBP
APOSPC32761-100G APOSPC32761-25G APOSPC32761-5G
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole 98%
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole
Formula: C₉H₇BrF₄O₂
MW: 303.048 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28040491
CAS Number: 220004-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR