4-Fluoro-2-methyl-6-nitroaniline 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC3258-1G PC3258-5G PC3258-10G
APOSPC3258-1GEA 15.8 GBP
APOSPC3258-1G APOSPC3258-5G APOSPC3258-10G
4-Fluoro-2-methyl-6-nitroaniline 97%
4-Fluoro-2-methyl-6-nitroaniline
Formula: C₇H₇FN₂O₂
Melting Pt: 98…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09152778
CAS Number: 147285-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR