Methyl 2-methyl-6-(trifluoromethyl)nicotinate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(Ethoxycarbonyl)-2-methyl-6-(trifluoromethyl)pyridine

PC31131-1G PC31131-5G PC31131-10G
APOSPC31131-1GEA 63 GBP
APOSPC31131-1G APOSPC31131-5G APOSPC31131-10G
Methyl 2-methyl-6-(trifluoromethyl)nicotinate
Methyl 2-methyl-6-(trifluoromethyl)nicotinate
Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01311989
CAS Number: 205582-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR