3,5-Bis(trifluoromethyl)phenol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: α,α,α,α',α',α'-Hexafluoro-3,5-xylenol

PC3112-100G PC3112-25G
APOSPC3112-100GEA 94.9 GBP
APOSPC3112-100G APOSPC3112-25G
3,5-Bis(trifluoromethyl)phenol 98%
3,5-Bis(trifluoromethyl)phenol
Formula: C₈H₄F₆O
Boiling Pt: 97…98 °C/50mmHg
Melting Pt: 20…21 °C
Density: 1.511 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR