2-Chloro-5-fluoroisonicotinic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC305019-1G
APOSPC305019-1GEA 17.9 GBP
APOSPC305019-1G
2-Chloro-5-fluoroisonicotinic acid
2-Chloro-5-fluoroisonicotinic acid
Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD04972381
CAS Number: 884494-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR