3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303569-1G PC303569-5G
APOSPC303569-1GEA 143 GBP
APOSPC303569-1G APOSPC303569-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid 98%
3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32640147
CAS Number: 2386483-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR