4-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303149-1G
APOSPC303149-1GEA 293 GBP
APOSPC303149-1G
4-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
4-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD28054194
CAS Number: 1706438-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR