5-Fluoro-2-(trifluoromethoxy)benzaldehyde 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC303002-10G PC303002-5G PC303002-1G
APOSPC303002-10GEA 179 GBP
APOSPC303002-10G APOSPC303002-5G APOSPC303002-1G
5-Fluoro-2-(trifluoromethoxy)benzaldehyde 97%
5-Fluoro-2-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11519331
CAS Number: 1092460-81-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR