3-Chloro-5-fluoro-4-methoxybenzyl bromide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC302950-10G PC302950-5G PC302950-1G
APOSPC302950-10GEA 293 GBP
APOSPC302950-10G APOSPC302950-5G APOSPC302950-1G
3-Chloro-5-fluoro-4-methoxybenzyl bromide
3-Chloro-5-fluoro-4-methoxybenzyl bromide
Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD04115868
CAS Number: 886497-36-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR