4,5-Difluoro-2-methoxybenzoyl chloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302855-10G PC302855-5G PC302855-1G
APOSPC302855-10GEA 204 GBP
APOSPC302855-10G APOSPC302855-5G APOSPC302855-1G
4,5-Difluoro-2-methoxybenzoyl chloride 97%
4,5-Difluoro-2-methoxybenzoyl chloride
Formula: C₈H₅ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06660197
CAS Number: 886498-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR