3-Fluoro-2-methoxybenzamide 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302840-1G PC302840-5G
APOSPC302840-1GEA 94.9 GBP
APOSPC302840-1G APOSPC302840-5G
3-Fluoro-2-methoxybenzamide 97%
3-Fluoro-2-methoxybenzamide
Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD06637313
CAS Number: 874830-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR