2-Fluoro-4-methoxybenzamide 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302803-10G PC302803-5G PC302803-1G
APOSPC302803-10GEA 235 GBP
APOSPC302803-10G APOSPC302803-5G APOSPC302803-1G
2-Fluoro-4-methoxybenzamide 97%
2-Fluoro-4-methoxybenzamide
Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD01117440
CAS Number: 874804-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR