4-Ethoxy-3,5-difluorobenzoic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC302749-10G PC302749-5G PC302749-1G PC302749-25G PC302749-100G
APOSPC302749-10GEA 204 GBP
APOSPC302749-10G APOSPC302749-5G APOSPC302749-1G APOSPC302749-25G APOSPC302749-100G
4-Ethoxy-3,5-difluorobenzoic acid 97%
4-Ethoxy-3,5-difluorobenzoic acid
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258683
CAS Number: 1017779-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR