2-Chloro-3-(trifluoromethyl)cinnamic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302639-5G PC302639-1G
APOSPC302639-5GEA 90.7 GBP
APOSPC302639-5G APOSPC302639-1G
2-Chloro-3-(trifluoromethyl)cinnamic acid 97%
2-Chloro-3-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03002851
CAS Number: 682805-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR