5-Fluoro-2-methoxybenzylamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (5-Fluoro-2-methoxyphenyl)methanamine

PC302621-1G PC302621-5G PC302621-10G PC302621-25G
APOSPC302621-1GEA 81.9 GBP
APOSPC302621-1G APOSPC302621-5G APOSPC302621-10G APOSPC302621-25G
5-Fluoro-2-methoxybenzylamine 97%
5-Fluoro-2-methoxybenzylamine
Formula: C₈H₁₀FNO
MW: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213625
CAS Number: 148870-38-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR