4-Fluoro-3-(trifluoromethoxy)benzamide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302594-1G PC302594-5G
APOSPC302594-1GEA 131 GBP
APOSPC302594-1G APOSPC302594-5G
4-Fluoro-3-(trifluoromethoxy)benzamide 98%
4-Fluoro-3-(trifluoromethoxy)benzamide
Formula: C₈H₅F₄NO₂
MW: 223.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772784
CAS Number: 1206593-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR