5-Fluoro-2-(trifluoromethoxy)aniline 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC302356-1G PC302356-5G PC302356-10G PC302356-25G
APOSPC302356-1GEA 15.8 GBP
APOSPC302356-1G APOSPC302356-5G APOSPC302356-10G APOSPC302356-25G
5-Fluoro-2-(trifluoromethoxy)aniline 98%
5-Fluoro-2-(trifluoromethoxy)aniline
Formula: C₇H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD16652514
CAS Number: 123572-63-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR