2-Methoxy-5-(trifluoromethoxy)benzonitrile 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302176-5G PC302176-1G PC302176-25G
APOSPC302176-5GEA 65.1 GBP
APOSPC302176-5G APOSPC302176-1G APOSPC302176-25G
2-Methoxy-5-(trifluoromethoxy)benzonitrile 97%
2-Methoxy-5-(trifluoromethoxy)benzonitrile
Formula: C₉H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD04115963
CAS Number: 886500-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR