4-Chloro-3-fluorobenzamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302107-25G PC302107-5G PC302107-10G
APOSPC302107-25GEA 315 GBP
APOSPC302107-25G APOSPC302107-5G APOSPC302107-10G
4-Chloro-3-fluorobenzamide
4-Chloro-3-fluorobenzamide
Formula: C₇H₅ClFNO
Storage Temperature: Ambient
CAS Number: 874781-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR