Dimethyl octafluoroadipate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC3015T-100G PC3015T-25G PC3015T-5G
APOSPC3015T-100GEA 248 GBP
APOSPC3015T-100G APOSPC3015T-25G APOSPC3015T-5G
Dimethyl octafluoroadipate 98%
Dimethyl octafluoroadipate
Formula: C₈H₆F₈O₄
Boiling Pt: 120 °C
Density: 1.547 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042267
CAS Number: 3107-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR