3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300781-1G PC300781-250MG PC300781-5G
APOSPC300781-1GEA 168 GBP
APOSPC300781-1G APOSPC300781-250MG APOSPC300781-5G
3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole
3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole
Formula: C₁₀H₇F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD26939177
CAS Number: 149739-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR