4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300758-1G PC300758-5G
APOSPC300758-1GEA 144 GBP
APOSPC300758-1G APOSPC300758-5G
4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide
4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide
Formula: C₁₆H₂₀ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD27995626
CAS Number: 1858255-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR